Yttrium(III)nitraatti-6 H2O 99,9

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Yttrium trinitrate hexahydrate

12898.06 12898.14 12898.22 12898.59
12898.06EA 87.1 EUR
12898.06 12898.14 12898.22 12898.59
Yttrium(III)nitraatti-6 H2O 99,9
Yttrium(III)nitraatti-6 H2O

Purity: 99.9% CAS No.: 13494-98-9 Molecular Formula: Y(NO3)3.6H2O Formula Weight: 383.01 (274.92anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,10107 UN No.: 1477 MDL No.: MFCD00149940

Kaava: N₃O₉Y·6H₂O
MW: 383.01 g/mol
Sulamispiste: 51.8
Tiheys: 2.682 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149940
CAS-numero: 13494-98-9
EINECS: 233-802-6
UN: 1477
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR