Rubidiumkloridi ≥99% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Rubidiumchloride

12892.14 12892.22 12892.06
12892.14EA 461 EUR
12892.14 12892.22 12892.06
Rubidiumkloridi ≥99% (metals basis)
Rubidiumkloridi

Purity: 99% CAS No.: 7791-11-9 Molecular Formula: RbCl Formula Weight: 120.92 Physical Form: Crystalline Density: 2.800 Refractive Index: 1.493 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 718° Boiling Point: 1390° Solubility: Soluble in water. Practically insoluble in alcohol Merck Reference: 14,8284 MDL No.: MFCD00011187


Testing for perchloric acid, source of rubidium, catalyst

Kaava: RbCl
MW: 120.92 g/mol
Kiehumispiste: 1390 °C
Sulamispiste: 718 °C
Tiheys: 2.800
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011187
CAS-numero: 7791-11-9
EINECS: 232-240-9
Merck Index: 14,8284

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR