Cesiumfluoridi ≥99% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

12885.14 12885.22
12885.14EA 177 EUR
12885.14 12885.22
Cesiumfluoridi ≥99% (metals basis)
Cesiumfluoridi

Purity: 99% CAS No.: 13400-13-0 Molecular Formula: CsF Formula Weight: 151.90 Physical Form: Crystalline Density: 4.115 Appearance: White Odour: Odourless pH: 6.5-7.5 Melting Point: 682° Boiling Point: 1251° UN No.: 2923 MDL No.: MFCD00010960

Kaava: CsF
Kiehumispiste: 1251 °C
Sulamispiste: 682 °C
Tiheys: 4,115
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010960
CAS-numero: 13400-13-0
EINECS: 236-487-3
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR