Bis(dibenzylideneacetone)palladium

Toimittaja: Thermo Fisher Scientific


Packaged under argon.

12764.01 12764.03 12764.06
12764.01EA 78.6 EUR
12764.01 12764.03 12764.06
Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium

CAS No.: 32005-36-0 Molecular Formula: C34H28O2Pd Formula Weight: 575.00 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Brown to purple Melting Point: 135° dec. Solubility: Slightly soluble in CH2Cl2, CHCl3 and benzene MDL No.: MFCD00051942


Applications: Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation

Kaava: C₃₄H₂₈O₂Pd
Sulamispiste: 135 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00051942
CAS-numero: 32005-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR