Lyijy(II)jodidi ≥99.9985% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

12724.14 12724.22
12724.14EA 342 EUR
12724.14 12724.22
Lyijy(II)jodidi ≥99.9985% (metals basis)
Lyijy(II)jodidi

Purity: 99.9985% CAS No.: 10101-63-0 Molecular Formula: I2Pb Formula Weight: 461.00 Physical Form: Powder Density: 6.16 Appearance: Yellow Odour: Odourless Melting Point: 402° Boiling Point: 954° Merck Reference: 14,5411 UN No.: 2291 MDL No.: MFCD00011163

Kaava: I₂Pb
Kiehumispiste: 954 °C
Sulamispiste: 402 °C
Tiheys: 6,16
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011163
CAS-numero: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 14,5411

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR