Kaliumbromidi ≥99% ACS

Toimittaja: Thermo Fisher Scientific
Varoitus

12612.18 12612.30 12612.A1 12612.A7
12612.18EA 65.5 EUR
12612.18 12612.30 12612.A1 12612.A7
Kaliumbromidi ≥99% ACS
Kaliumbromidi

Purity: 99% min CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Physical Form: Crystalline Density: 2.75 Refractive Index: 1.559 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 730° Boiling Point: 1435° Solubility: Freely soluble in water. Soluble in alcohol, glycerol Merck Reference: 14,7618 MDL No.: MFCD00011358


Manufacturing of photographic papers, process engraving and lithography, spectroscopy, as an analytical lab reagent

Kaava: KBr
Kiehumispiste: 1435 °C
Sulamispiste: 730 °C
Tiheys: 2,75
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011358
CAS-numero: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR