Indium(III)oksidi ≥99.99% (metals basis)

Toimittaja: Thermo Fisher Scientific

12544.09 12544.18 12544.30
12544.09EA 278 EUR
12544.09 12544.18 12544.30
Indium(III)oksidi ≥99.99% (metals basis)
Indium(III)oksidi

Purity: 99.99% CAS No.: 1312-43-2 Molecular Formula: In2O3 Formula Weight: 277.64 Physical Form: -325 Mesh Powder Density: 7.18 Melting Point: 850° volat. Merck Reference: 14,4952 MDL No.: MFCD00011060

Kaava: In₂O₃
Sulamispiste: 850 °C
Tiheys: 7,18
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011060
CAS-numero: 1312-43-2
EINECS: 215-193-9
Merck Index: 14,4952

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR