Yttrium ≥99.6% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Toimittaja: Thermo Fisher Scientific

12442.FF 12442.FI
12442.FFEA 304 EUR
12442.FF 12442.FI
Yttrium ≥99.6% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Yttrium

Packaged in mylar under argon


Purity: 99.9% CAS No.: 7440-65-5 Physical Form: ≈0.28g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011468

Kaava: Y
MW: 88.91 g/mol
Kiehumispiste: 2927
Sulamispiste: 1525
Tiheys: 4.472 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011468
CAS-numero: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR