Skandium 99,9 REO basis, foil, Thickness: 0.025 mm (0.001 in)

Toimittaja: Thermo Fisher Scientific
Varoitus


Argon_filled

12426.FF 12426.FI
12426.FFEA 681 EUR
12426.FF 12426.FI
Skandium 99,9 REO basis, foil, Thickness: 0.025 mm (0.001 in)
Skandium

Purity: 99.9% CAS No.: 7440-20-2 Physical Form: ≈0.05g/25x25mm, Packaged under argon Appearance: Grey Odour: Odourless UN No.: 3178 MDL No.: MFCD00016323

Kaava: Sc
MW: 44.96 g/mol
Kiehumispiste: 2730
Sulamispiste: 1541
Tiheys: 2.989 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016323
CAS-numero: 7440-20-2
EINECS: 231-129-2
UN: 3178
ADR: 4.1 III
Merck Index: 12,08537

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR