Kalsiumkarbonaatti ≥98%

Toimittaja: Thermo Fisher Scientific

12365.36 12365.A3
12365.36EA 107 EUR
12365.36 12365.A3
Kalsiumkarbonaatti ≥98%
Kalsiumkarbonaatti

Purity: 98% CAS No.: 471-34-1 Molecular Formula: CaCO3 Formula Weight: 100.09 Physical Form: Powder Density: 2.930 Refractive Index: 1.6583 Appearance: White Odour: Odourless Melting Point: 800° dec. Solubility: Practically insoluble in water. Soluble in diluted acids Merck Reference: 14,1657 MDL No.: MFCD00010906


Manufacture of paint, rubber, plastics, etc. In food, cosmetics, and pharmaceuticals. As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer

Kaava: CaCO₃
Sulamispiste: 800 °C (haj. lämpötila)
Tiheys: 2,930
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010906
CAS-numero: 471-34-1
EINECS: 207-439-9
Merck Index: 14,1657

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR