Seleenidioksidi ≥99.4% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

12358.22 12358.36 12358.A1
12358.22EA 147 EUR
12358.22 12358.36 12358.A1
Seleenidioksidi ≥99.4% (metals basis)
Seleenidioksidi

Purity: 99.4% CAS No.: 7446-08-4 Molecular Formula: O2Se Formula Weight: 110.96 Physical Form: Powder Density: 3.95 Appearance: White to pink Odour: Odourless Melting Point: 340-350° subl. Solubility: Partially soluble in water, methanol, ethanol, acetone. Soluble in concentrated H2SO4 Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562


As an oxidizing agent, in manufacturing of other selenium compounds

Kaava: SeO₂
MW: 110.96 g/mol
Sulamispiste: 340…350 °C (subl.)
Tiheys: 3.95
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003562
CAS-numero: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1 II
Merck Index: 14,8434

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR