Anisoli 99%, Reagent Grade

Toimittaja: Honeywell Chemicals
Varoitus

Synonyms: Methyl phenyl ether, Methoxybenzene

123226-1L 123226-2.5L
123226-1LEA 93.8 EUR
123226-1L 123226-2.5L
Anisoli 99%, Reagent Grade
Anisoli

  • ≥95% for general laboratory use.

Kaava: C₇H₈O
MW: 108.14 g/mol
Kiehumispiste: 154 °C (1.013 hPa)
Sulamispiste: –37 °C
Tiheys: 1.00 g/cm³ (20 °C)
Leimahduspiste: 43 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008354
CAS-numero: 100-66-3
EINECS: 202-876-1
UN: 2222
ADR: 3 III
Merck Index: 13,00672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d32206d4bb9dbd12360d11d39b31237d54a8dcf8&vid=286a07a5e34a1ec55ff6339785af95d9a4326825&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Vapor Density 3.7 (vs air)
Assay (GC) Min. 98.5 %
Identity (IR) complying
Vapor Pressure 10 mmHg ( 42.2 °C)
Autoignition Temperature 887 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR