Magnesiumkloridi, vedetön 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

12315.A1 12315.A7 12315.C4
12315.A1EA 142 EUR
12315.A1 12315.A7 12315.C4
Magnesiumkloridi, vedetön 99%
Magnesiumkloridi

Purity: 99% CAS No.: 7786-30-3 Molecular Formula: MgCl2 Formula Weight: 95.22 Physical Form: Crystalline Density: 2.32 Appearance: White to yellow Odour: Odourless Melting Point: 714° Solubility: Soluble in water with evolution of heat Merck Reference: 14,5662 MDL No.: MFCD00011106


Catalyst, flocculating agent, various magnesium cements, ceramics, analytical reagent

Kaava: MgCl₂
Kiehumispiste: 1412 °C (1013 hPa)
Sulamispiste: 714 °C
Tiheys: 2,32
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011106
CAS-numero: 7786-30-3
EINECS: 232-094-6
Merck Index: 14,5662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR