Vismutti(III)oksidi ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

12230.30 12230.A1 12230.A7
12230.30EA 151 EUR
12230.30 12230.A1 12230.A7
Vismutti(III)oksidi ≥99%
Vismutti(III)oksidi

Purity: 99% CAS No.: 1304-76-3 Molecular Formula: Bi2O3 Formula Weight: 465.96 Physical Form: Powder Density: 8.9 Appearance: Yellow Melting Point: 817° Boiling Point: ca 1890° Solubility: Practically insoluble in water. Soluble in acids Merck Reference: 14,1273 MDL No.: MFCD00003462


In disinfectants, magnets, glass, rubber, vulcanization, in fireproofing papers and polymers, and in catalysts

Kaava: BiO₃
Kiehumispiste: ∼1890 °C
Sulamispiste: 817 °C
Tiheys: 8,9
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003462
CAS-numero: 1304-76-3
EINECS: 215-134-7
Merck Index: 14,1273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR