Natriummolybdaatti (VI) dihydraatii 99.5-103.0% ACS

Toimittaja: Thermo Fisher Scientific

12214.22 12214.36
12214.22EA 144 EUR
12214.22 12214.36
Natriummolybdaatti (VI) dihydraatii 99.5-103.0% ACS
Natriummolybdaatti (VI) dihydraatii

Purity: 99.5-103.0% CAS No.: 10102-40-6 Molecular Formula: MoNa2O4.2H2O Formula Weight: 241.95 (205.92anhy) Physical Form: Powder Density: 2.71 Appearance: White Odour: Odourless Melting Point: 100° -2H2O Solubility: Soluble in water Merck Reference: 14,8645 MDL No.: MFCD00149170

Kaava: Na₂MoO₄·2H₂O
Sulamispiste: 100 °C (-2H₂O)
Tiheys: 2,71
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149170
CAS-numero: 10102-40-6
EINECS: 231-551-7
Merck Index: 14,8645

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR