Iridium(III) bromide hydrate (≥35,9% Ir)

Toimittaja: Thermo Fisher Scientific

12212.03 12212.06
12212.03EA 695 EUR
12212.03 12212.06
Iridium(III) bromide hydrate (≥35,9% Ir)
Iridium(III) bromide hydrate

Molecular Formula: IrBr3.xH2O Formula Weight: 431.91(anhy) Physical Form: Crystalline Melting Point: 100° -3H2O MDL No.: MFCD00049466

Kaava: IrBr₃·xH₂O
Sulamispiste: 100 °C (-3H₂O)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049466
EINECS: 233-174-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR