Agaroosi molekyylibiologiaan Type 1, Millipore®

Toimittaja: Merck Millipore Calbiochem

121853-100 121853-500GM
121853-100EA 407 EUR
121853-100 121853-500GM
Agaroosi molekyylibiologiaan Type 1, Millipore®
Agaroosi

It is suitable for a wide range of nucleic acid electrophoresis and immunoelectrophoresis applications.


Synonyms: Agarose 10; Agarose 15; Agarose 25; Agarose IEF.

Sulamispiste: 87…89 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081294
CAS-numero: 9012-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Gel strength ≥1200 g/cm²
Gelling range (1,5%) 36 - 39 °C
EEO (-mr) ≤0,10
Sulphate ≤0,15%
DNase, RNase, protease None detected
Storage Store at 15 to 30 °C, protect from moisture


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR