Sodium hexachloroosmate(IV) dihydrate Os ≥38,7%

Toimittaja: Thermo Fisher Scientific
Varoitus

12176.01 12176.03 12176.06
12176.01EA 156 EUR
12176.01 12176.03 12176.06
Sodium hexachloroosmate(IV) dihydrate Os ≥38,7%
Sodium hexachloroosmate(IV) dihydrate

CAS No.: 1307-81-9 Molecular Formula: Cl6Na2Os.2H2O Formula Weight: 484.93 (448.90anhy) Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Appearance: Red to brown Odour: Odourless MDL No.: MFCD00014234

Kaava: Cl₆Na₂Os·2H₂O
Kiehumispiste: 308 °C (1013 hPa)
Sulamispiste: 389…393 °C
Tiheys: 3,59 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00014234
CAS-numero: 1307-81-9
EINECS: 215-152-5

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR