PIG® SUPER absorbent socks
Toimittaja: NEW PIG
PIG210 PIG211 PIG214
121-0537EA 149 EUR
121-0537 NEWPPIG211 NEWPPIG214
PIG® SUPER absorbent socks
Imeytysaineet ja imeytysliinat
PIG® SUPER Absorbent Sock is great for high-volume machine leaks. Soaks up nearly twice as much liquid as other maintenance socks.
  • Extra-thirsty, formable socks hug corners and walls, surround higher-volume machine leaks and contain spills to keep workers safe
  • Cellulose filler is highly absorbent for big leaks and spills; super absorbency means that you replace them less often
  • Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water; not recommended for