PIG® spill kit in see-thru bag
Toimittaja: NEW PIG
KIT274 KIT367 KIT470
121-0508EA 94 EUR
121-0508 121-0518 121-0564
PIG® spill kit in see-thru bag
Imeytysaineet ja imeytysliinat
Compact, easy-to-inventory bag kit with zipper is perfect for stocking a fleet.
  • Compact kit with zipper closure and carry handle is ideal for outfitting a vehicle fleet
  • PIG socks stop spreading spills; PIG mat absorbs quickly
  • Temporary disposal bags help make cleanup easier
  • Bold Spill Kit lettering for quick identification
The transparent bag lets you check supply levels at a glance whilst keeping contents clean and dry.
Varoitus: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of yourMore Product Information