VWR®, Absorbents - Mats, Rolls and Socks, for Chemical Based Spills
Toimittaja: VWR Collection
121-0142EA 91.6 EUR
121-0142 121-0145 121-0139
VWR®, Absorbents - Mats, Rolls and Socks, for Chemical Based Spills
Imeytysaineet ja imeytysliinat
VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
  • Absorbs a wide range of chemicals, including acids and caustics
  • Chemically inert: Product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid
  • Green colour alerts that product is being used for hazardous liquids
  • Green colour allows user to identify and separate hazardous waste for disposal, resulting in lower disposal costs
  • Cannot be left outdoors as they will absorb water