Bottles, narrow neck, round, low particulate, HDPE, IP2, with screw caps, Nalgene®
Toimittaja: Thermo Fisher Scientific
2099-0001 2099-0002 2099-0004 2099-0008 2099-0010 2099-0016 2099-0032
119-4411EA 163 EUR
119-4411 119-4412 119-4413 NALG2099-0008 119-4417 119-4415 119-4416
Bottles, narrow neck, round, low particulate, HDPE, IP2, with screw caps, Nalgene®
Pullot Muovipullot
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Screw cap supplied assembled on the bottle
  • Leakproof
Utilize Nalgene™ Narrow-Mouth HDPE IP2 Bottles as inner containers for designing, assembling, and certifying combination packaging. These high density polyethylene bottles are safe, economical, and designed to resist splitting and puncturing from shipping shocks.
Varoitus: *Tällä sivulla olevat tuotteet eivät kuulu EU -direktiivin 2019/904 soveltamisalaan.