Potassium hexachloroiridate(III) hydrate (≥32,8% Ir)

Toimittaja: Thermo Fisher Scientific
Varoitus

11887.03 11887.06 11887.14
11887.03EA 639 EUR
11887.03 11887.06 11887.14
Potassium hexachloroiridate(III) hydrate (≥32,8% Ir)
Potassium hexachloroiridate(III) hydrate

Molecular Formula: K3IrCl6.xH2O Formula Weight: 522.22(anhy) Physical Form: Crystalline MDL No.: MFCD00038699

Kaava: K₃IrCl₆·xH₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00038699
EINECS: 237-854-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR