Indium(III) chloride hydrate ≥99.99% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

11859.09 11859.18 11859.30
11859.09EA 193 EUR
11859.09 11859.18 11859.30
Indium(III) chloride hydrate ≥99.99% (metals basis)
Indium(III) chloride hydrate

Purity: 99.99% CAS No.: 143983-91-9 Molecular Formula: InCl3.xH2O Formula Weight: 221.18(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Solubility: Soluble in water Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058

Kaava: Cl₃In·H₂O
MW: 221.18 g/mol (anhy)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149744
CAS-numero: 143983-91-9
EINECS: 233-043-0
UN: 3260
ADR: 8 II
Merck Index: 14,4958

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR