Hafnium(IV)kloridi ≥98% (metals basis excluding Zr) (max. 2.7% Zr)

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Hafnium tetrachloride

11834.14 11834.22 11834.88
11834.14EA 168 EUR
11834.14 11834.22 11834.88
Hafnium(IV)kloridi ≥98% (metals basis excluding Zr) (max. 2.7% Zr)
Hafnium(IV)kloridi

Purity: 98+% CAS No.: 13499-05-3 Molecular Formula: Cl4Hf Formula Weight: 320.30 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White Odour: Irritating Melting Point: 320° subl. Merck Reference: 14,4588 UN No.: 3260 MDL No.: MFCD00011034

Kaava: Cl₄Hf
MW: 320.3 g/mol
Sulamispiste: 320 °C (subl.)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011034
CAS-numero: 13499-05-3
EINECS: 236-826-5
UN: 3260
ADR: 8 II
Merck Index: 14,4588

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR