Sodium hexachlororhodate (III) dodecahydrate (17,1% Rh)

Toimittaja: Thermo Fisher Scientific
Varoitus

11809.03 11809.06
11809.03EA 1070 EUR
11809.03 11809.06
Sodium hexachlororhodate (III) dodecahydrate (17,1% Rh)
Sodium hexachlororhodate (III) dodecahydrate

CAS No.: 14972-70-4 Molecular Formula: Na3RhCl6.12H2O Formula Weight: 600.77 (384.59anhy) Physical Form: Crystalline Melting Point: 900° dec. MDL No.: MFCD00003509

Kaava: Cl₆Na₃Rh·12H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003509
CAS-numero: 14972-70-4
EINECS: 239-047-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR