Gas detection tubes for chlorine dioxide
Toimittaja: Dräger
8103491
118-0355EA 177 EUR
118-0355
Gas detection tubes for chlorine dioxide
Kaasuilmasinputket
To measure the 'spot concentrations' of chlorine dioxide with an Accuro® pump. Uses include determination of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces. The following values are not displayed:
  • 1 ppm Cl₂, 10 ppm H₂S, 1 ppm SO₂
  • 10 ppm methyl mercaptan
  • 1 ppm bromide is not displayed at a stroke count of n = 10, at n = 30 there is a discoloration of approx. 10 mm
One stroke is equivalent to a volume of 100 ±5 ml of gas passingMore Product Information