Gas detection tubes for benzene
Toimittaja: Dräger
8101231 6728071 8101741 6718801 8103691
151-0169EA 165 EUR
151-0169 118-0289 118-0317 DRAE6718801 DRAE8103691
Gas detection tubes for benzene
Kaasuilmasinputket
To measure the 'spot concentrations' of carbon dioxide for use with an Accuro® pump. Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
  • Benzene 5/a and 15/a with prelayer retains toluene and xylene causing a reddish brown discolouration
  • If xylene and toluene levels are too high, the entire prelayer up to the indicating layer is descoloured making benzene mesurement impossible
  • Petroleum