Platina 1% on gamma alumina, jauhe, reduced

Toimittaja: Thermo Fisher Scientific

11797.14 11797.22 11797.06
11797.14EA 779 EUR
11797.14 11797.22 11797.06
Platina 1% on gamma alumina, jauhe, reduced
Platina

CAS No.: 7440-06-4 Molecular Formula: 300m2/g MDL No.: MFCD00011179

Kaava: Pt
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011179
CAS-numero: 7440-06-4

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR