Hopea(I)asetaatti, vedetön 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

11660.14 11660.22 11660.A1
11660.14EA 356 EUR
11660.14 11660.22 11660.A1
Hopea(I)asetaatti, vedetön 99%
Hopea(I)asetaatti

Purity: 99% CAS No.: 563-63-3 Molecular Formula: C2H3AgO2 Formula Weight: 166.92 Physical Form: Crystalline Density: 3.26 Appearance: White to grey Odour: Odourless Melting Point: dec. Solubility: Soluble in dilute HNO3. Water solubility increases with temperature Merck Reference: 14,8505 UN No.: 3077 MDL No.: MFCD00012446

Kaava: C₂H₃AgO₂
Sulamispiste: Dec.
Tiheys: 3,26
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012446
CAS-numero: 563-63-3
EINECS: 209-254-9
UN: 3077
ADR: 9,III
Merck Index: 14,8505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR