Magnesiumperkloraatti, anhydrous ACS

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Anhydrone

11636.22 11636.36
11636.22EA 205 EUR
11636.22 11636.36
Magnesiumperkloraatti, anhydrous ACS
Magnesiumperkloraatti

CAS No.: 10034-81-8 Molecular Formula: Mg(ClO4)2 Formula Weight: 223.23 Physical Form: Granular Density: 2.21 Sensitivity: Hygroscopic Merck Reference: 14,5678 UN No.: 1475 MDL No.: MFCD00011102


Desiccant. Drying agent for gases

Kaava: Cl₂MgO₈
MW: 223.21 g/mol
Sulamispiste: 251 °C (haj. lämpötila)
Tiheys: 2.21
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011102
CAS-numero: 10034-81-8
EINECS: 233-108-3
UN: 1475
ADR: 5.1 II
Merck Index: 14,5678

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR