Kaliumdivetyortofosfaatti ≥99.0% ACS

Toimittaja: Thermo Fisher Scientific
Varoitus

11594.30 11594.A1 11594.36
11594.30EA 115 EUR
11594.30 11594.A1 11594.36
Kaliumdivetyortofosfaatti ≥99.0% ACS
Kaliumdivetyortofosfaatti

Purity: 99.0% min CAS No.: 7778-77-0 Molecular Formula: H2KO4P Formula Weight: 136.09 Physical Form: Crystalline Density: 2.338 Appearance: White Melting Point: 253° Solubility: Slightly soluble in water. Insoluble in alcohol Merck Reference: 14,7659 MDL No.: MFCD00011401


In buffers for pH determination.

Kaava: KH₂PO₄
Kiehumispiste: ∼100 °C (1013 hPa)
Sulamispiste: 253 °C
Tiheys: 2,338
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011401
CAS-numero: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 14,7659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR