Antimonitrioksidi ≥99.6%

Toimittaja: Thermo Fisher Scientific
Varoitus

11579.18 11579.36 11579.A3
11579.18EA 82.1 EUR
11579.18 11579.36 11579.A3
Antimonitrioksidi ≥99.6%
Antimonitrioksidi

Purity: 99.6% min CAS No.: 1309-64-4 Molecular Formula: Sb2O3 Formula Weight: 291.50 Physical Form: 1.1-1.8 Micron Powder Density: 5.67 Appearance: White Odour: Odourless Melting Point: 656° Boiling Point: 1550° subl. Solubility: Slightly soluble in water, Slowly dissolves in warm HCl or warm KOH solutions Merck Reference: 14,711 MDL No.: MFCD00011214

Kaava: O₃Sb₂
Kiehumispiste: 1550 °C (subl.)
Sulamispiste: 656 °C
Tiheys: 5,67
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011214
CAS-numero: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1,III
Merck Index: 14,711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR