VWR®, Suojapaperi
Toimittaja: VWR Collection
115-9220EA 77.4 EUR
115-9220 115-9221
VWR®, Suojapaperi
Suojapaperit ja pintasuojukset Pöydänsuojukset
High quality, smooth, absorbent paper that quickly absorbs liquid spills with a thick, laminated polyethylene layer that prevents seeping onto the work surface.
  • Retains leaked reagents in radiochemical laboratories preventing radioactive contamination
  • Enables recovery of expensive leaked materials
  • Breakage protection for hard surfaces
  • Absorption layer for removing water or solvent from desiccant chambers
  • Impermeable protective material for lining animal cages
Absorbance rate: 115-9220 and 115-9221: 500 ml/m²
Absorbance rate: 115-0673: 750 ml/m²