Conical earth bonding point cord
Toimittaja: DESCO
60474
115-3069EA 20.5 EUR
115-3069
Conical earth bonding point cord
Maadoittajat Maadoituskaapelit
This cord with low profile helps keep employees from tripping on cord connection.
  • 10 mm snap stud easily snaps to mat socket, flush to mat surface
  • Extra ring terminal provided for customising cord length
  • Ring terminal at end of cord allows easy attachment to building ground
Stud has threaded hole; 6-32×6,35 mm flat head screw and countersunk washer are provided which allows ground cord to be bolted to mat; keeps cord from disconnecting; ensures proper grounding.
Toimitustiedot: Includes 1 Megohm resistor.