Cleanroom wipes, pre-saturated, PROSAT® Pi
Toimittaja: CONTEC
PS-314-BPR PS-314IR-BPR
115-2944EA 1030 EUR
115-2944 115-2945
Cleanroom wipes, pre-saturated, PROSAT® Pi
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Esikyllästetyt puhdastilan pyyhkeet
Hydroentangled split microfibre wipes pre-saturated with 70% IPA and 30% DI water.
  • Greater surface coverage means less resource consumption including time, labour and materials
  • Pre-saturated with 70% IPA / 30% DI water; controls saturation levels which greatly reduce solvent usage, VOCs and waste
  • Non woven split microfibre increases cleaning effectiveness and contaminant pick-up
PROSAT® Pi wipes are designed to help life science manufacturers control contamination and ultimately maintain cGMP compliance while consuming less resources. The wipes areMore Product Information