Puhdistusliinat, yleiskäyttöön, Multitex®
Toimittaja: Zellstoff-Vertriebs-GmbH & Co. KG
10149B 10149A 11149-01
115-2798EA 147 EUR
115-2798 115-0567 ZVGA11149-01
Puhdistusliinat, yleiskäyttöön, Multitex®
Pyyhkeet, liinat ja sienet Kertakäyttöliinat ja -pyyhkeet Paperiliinat yleiskäyttöön
These high-quality viscose fleece wipes with textile characteristics are suitable for wet and dry cleaning. The wipes are very soft and yet lint-free, they can absorb large amounts of liquid, remain tear-resistant and meet food safety requirements.
  • Solvent resistant and fibre-free
  • Highly absorbent
  • High tear resistance, even when wet
Ideal for use in sensitive work areas, such as precision mechanics, electrical and automotive industries, and in paint shops.