Cleanroom wipes, presaturated, PROSAT® Theta™
Toimittaja: CONTEC
PSC20006-BPR
115-2783EA 1210 EUR
115-2783
Cleanroom wipes, presaturated, PROSAT® Theta™
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Esikyllästetyt puhdastilan pyyhkeet
These PROSAT® wipes are manufactured from a lightweight hydroentangled cellulose/polyester and saturated with a solution of 70% USP grade IPA and 30% deionized water.
  • Suitable for ISO Class 5-8 environments
  • Nonwoven material is low in particles and fibres, making an excellent general purpose wipe
  • Highly sorbent with good wet strength
  • Presaturated wipe reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels
A cost effective wipe ideal for transfer disinfection and general cleaning, PROSAT® Theta™ wipesMore Product Information