Sterile meltblown polypropylene wipes, PROSAT®
Toimittaja: CONTEC
PS-911EB-BPR PSPP0039-BPR
115-2772EA 1530 EUR
115-2772 115-2815
Sterile meltblown polypropylene wipes, PROSAT®
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Esikyllästetyt puhdastilan pyyhkeet
Presaturated with 70% USP grade IPA and 30% DI water.
  • Suitable for Grade A/B or ISO Class 5-8 environments
  • Validated sterile per gamma irradiation
  • Meltblown polypropylene fabric is exceptionally clean and free from additives
  • Low in particles and fibres, ideal for product contact areas
  • Microsized filaments provide exceptional particle removal
  • Especially suitable for the metered release of 70% alcohol
  • Reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels
PROSAT® Sterile Polypropylene wipes areMore Product Information