Food and hygiene wiping paper, centrefeed roll, WypAll® Reach™
Toimittaja: KIMBERLY CLARK
6222 6223
115-2529EA 64.2 EUR
115-2529 115-2531
Food and hygiene wiping paper, centrefeed roll, WypAll® Reach™
Pyyhkeet, liinat ja sienet Kertakäyttöliinat ja -pyyhkeet Paperiliinat yleiskäyttöön
WypAll® L10 centrefeed roll, designed for light-duty hygiene wiping and perfect for tasks like cleaning food preparation and service surfaces. Compatible with the portable WypAll® Reach™ dispenser (115-2528) which ensures rolls remain enclosed to help prevent against transmission of food-borne bacteria.
  • Each WypAll® L10 centrefeed roll contains 430 embossed wiping paper sheets
  • Designed specifically for the WypAll® Reach™ wiping paper dispenser, using the red dispensing nozzle
  • Wiping rolls remain clean and enclosed to help prevent against transmission of