Regenerating hand cream, Stokoderm® Grip PURE
Toimittaja: Deb-STOKO
SGP100ML SGP1L
115-1986EA 6.8 EUR
115-1986 115-2359
Regenerating hand cream, Stokoderm® Grip PURE
Käsienpesu- ja desinfektioaineet, hoitovoiteet Voiteet Hand Creams
Versatile skin protection product (O/W emulsion) for sensitive skin exposed to changing and non specific stresses. Can also be used under gloves. The product is effective for both purely oil-based and water-miscible or aqueous industrial substances. Ideal for activities in changing working areas.
  • Ingredients include an astringent active substance, wax-like agents and emollients
  • Free from classical preservatives, perfume, colourants and silicone
  • With gentle, moisture-binding ingredients
  • Can also be applied to the face due to good compatibility with mucosal