Puhdastilan pyyhkeet, lattialiinat, steriilit, AlphaLite® II
Toimittaja: Texwipe
TX3424
115-1893EA 1150 EUR
115-1893
Puhdastilan pyyhkeet, lattialiinat, steriilit, AlphaLite® II
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Steriilit puhdastilan pyyhkeet
Cut-edge polyester single ply wipe that is ideal for floor cleaning and for applying cleaning solutions and disinfectants on floors.
  • Large size for improved application on floor surfaces
  • Size is compatible with most mopping systems
  • Improved contamination control
Suitable for use in Class 100, ISO 5 and Class 1000, ISO 6 environments.
Pakkaus: Packaging is adapted for sterile and highly critical environments.