Puhdastilan pyyhkeet, Super Polx® SWSE
Toimittaja: BERKSHIRE
SPSWSE.0909.10 SPSWSE.0909B
115-1786EA 789 EUR
115-1786 115-1787
Puhdastilan pyyhkeet, Super Polx® SWSE
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Kuivat puhdastilan pyyhkeet
This sealed edge cleanroom wipe made of 100% continuous filament double knitted polyester is excellent for general wiping and disinfecting of critical process areas and machines.
  • Strong, tear and abrasion resistant
  • Excellent absorbency
  • Low particles and fibres
  • Sterilisable by autoclave, Gamma or Beta
  • Laundered and packaged in an ISO 4 (Class 10) cleanroom; lot tracing on each pack
Ideal for economic high volume cleaning and wipe-downs.
Suitable for use in Class 1, ISO 3, M1.5 and above cleanrooms.
Varoitus: Data accurate at time ofMore Product Information