Puhdastilan pyyhkeet, esikyllästetty, steriilit, Vertex®
Toimittaja: Texwipe
TX3042P TX3049P TX3044P
115-1412EA 74.6 EUR
115-1412 115-1410 115-1890
Puhdastilan pyyhkeet, esikyllästetty, steriilit, Vertex®
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Esikyllästetyt puhdastilan pyyhkeet
These 100% continuous filament, polyester knit fabric wipes are made from virgin stock and processed on Texwipe’s Vertex® manufacturing system. The Vertex® manufacturing process cleans and converts wipes in an integrated, hands-free micro environment. The wipes are manufactured in an ISO class 3 environment with a small physical footprint which provides for a more sustainable process.
  • Sealed edge wipes
  • Special design provides a very soft feel and high sorptive capacity
  • Ideal for use with delicate surfaces or applications
  • Wipes do not contribute cytotoxic