VWR®, Floor Mat Frames for PureStep Mats
Toimittaja: VWR Collection
115-1296EA 202 EUR
115-1296 115-1287 115-1288 115-1289 115-1290 115-1291 115-1292 115-2471
VWR®, Floor Mat Frames for PureStep Mats
Matot Mat Frames
These frames are designed to be used with PureStep mats to create a system, that is ideal for any critical area, and does not allow direct adhesion to the flooring surface. The frames are constructed of a hard, rigid, polystyrene base material, combined with a non skid backing, that holds the mats in place on almost any surface.
  • Protects floors from adhesive residue
  • Reduces the risk of slips and falls
  • Non slip parts made from polymer coated, polyester textile
  • Naturally antimicrobial, mould and mildew resistant
Tilaustiedot: Adhesive mats are sold separately.