Surface protection papers, LabSorb
Toimittaja: Ahlstrom-Munksjö
146298 146313 146317 146289 146299 146306 146287 146318 146297 146314 146288 1.601.920.050
115-0575EA 350 EUR
115-0575 115-0578 115-0577 115-0576 BINZ146299 BINZ146306 BINZ146287 BINZ146318 BINZ146297 BINZ146314 BINZ146288 BINZ1.601.920.050
Surface protection papers, LabSorb
Suojapaperit ja pintasuojukset Pöydänsuojukset
Double-layered LabSorb papers protect benches and working areas from spilled chemicals, corrosive materials, as well as toxic, infectious or radioactive substances. Spills are quickly absorbed into the surface layer of cellulose material and prevented from penetrating to the surface of the work bench by a thin coating of polyethylene that serves as a carrier for the absorbent layer.
  • Top layer consists of highly absorbent cellulose to quickly capture any spills
  • Bottom layer is an impermeable polyethylene coating that prevents contamination of the covered