Puhdastilan pyyhkeet, pyyhkeet ja sienet, UltraSOLV®
Toimittaja: FOAMTEC
HT4790 HT4754 HT4794-25 HT4790-5
115-0283EA 7.7 EUR
115-0283 115-0282 FOAMHT4794-25 115-0428
Puhdastilan pyyhkeet, pyyhkeet ja sienet, UltraSOLV®
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Kuivat puhdastilan pyyhkeet
These highly absorbent wipes are constructed from a naturally reticulated, hydrophilic, polyurethane foam.
  • High abrasion resistance and conformable characteristics of UltraSOLV® allow it to clean rough, coarse and bead blasted surfaces that will snag traditional polyester wipes
  • Low levels of particle generation, ionics, organics, lot-to-lot consistency and fibre-free construction
  • Unsurpassed contamination removal properties
  • Sponge-like properties allow for excellent pick up and release of fluids
  • Excellent static dissipative and tribocharging properties