Indium ≥99.998% (metals basis), lanka, Puratronic®, Ø 0.5 mm (0.02 in)

Toimittaja: Thermo Fisher Scientific

11463.G3 11463.G6 11463.G9
11463.G3EA 401 EUR
11463.G3 11463.G6 11463.G9
Indium ≥99.998% (metals basis), lanka, Puratronic®, Ø 0.5 mm (0.02 in)
Indium

Purity: 99.998+% CAS No.: 7440-74-6 Physical Form: ≈1.43g/m Odour: Odourless MDL No.: MFCD00134048

Kaava: In
Kiehumispiste: 2000 °C (1013 hPa)
Sulamispiste: 156 °C
Tiheys: 7,3 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134048
CAS-numero: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR