Tina ≥99.998% (metals basis), folio, Puratronic®, Thickness: 0.1 mm (0.004 in)

Toimittaja: Thermo Fisher Scientific

11460.FIEA 163 EUR
11460.FI 11460.GH
Tina ≥99.998% (metals basis), folio, Puratronic®, Thickness: 0.1 mm (0.004 in)
Tina

Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈1.8g/50x50mm Odour: Odourless MDL No.: MFCD00133862

Kaava: Sn
Kiehumispiste: 2270 °C (1013 hPa)
Sulamispiste: 232 °C
Tiheys: 7,31 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00133862
CAS-numero: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR