Pouch masks with nylon headband, non sterile, HM4, PUREZERO®
Toimittaja: Halyard
CLN3941BBG
114-9601EA 165 EUR
114-9601
Pouch masks with nylon headband, non sterile, HM4, PUREZERO®
Maskit Puhdastilamaskit
The HM4 Pouch Mask is a three-layer mask with nylon headband with an expanded breathing chamber that provides high particle, bacterial, and viral filtration efficiency. The mask construction consists of low-linting materials and a polyester foam nose band.
  • Three-layer mask
  • Nylon headband
  • Designed for industrial and cleanroom use
  • Recommended for ISO Class 4 or higher and Grade B/C/D cleanrooms
  • Exclusive SO SOFT biocomponent technology inner lining for comfort
Cleanroom efficiency depends on effective risk management. Ensure masks are up to the taskMore Product Information